Assay Details
Target Gene Details
Entrez Gene ID: | 328108 |
Gene Name: | family with sequence similarity 179, member B |
Gene Aliases: |
A430041B07Rik, BC062107, C130034K06, mKIAA0423 |
Location: |
Chr.12:64965742-65022573 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam179b | NM_177805.3 | 4 | 2724 | NP_808473.2 |
| AK039995.1 | 5 | 2435 | BAC30493.1 | |
| AK132890.1 | 2 | 221 | ||
| AK141238.1 | 4 | 2664 | ||
| AK172948.1 | 4 | 2652 | BAD32226.1 | |
| BC157958.1 | 4 | 2526 | ||
| BC158008.1 | 4 | 2526 | ||
| BC172117.1 | 4 | 2526 |
More Information
Set Membership: |
|

Genomic Map