Assay Details
Target Gene Details
Entrez Gene ID: | 80837 |
Gene Name: | ras homolog gene family, member J |
Gene Aliases: |
1110005O19Rik, AW210585, Arhj, TC10L, TCL |
Location: |
Chr.12:75308313-75401455 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhoj | NM_023275.2 | 5 | 1454 | NP_075764.1 |
| AB060651.1 | 5 | 1359 | BAB91069.1 | |
| AF309564.1 | 5 | 1313 | AAL09441.1 | |
| AK003482.1 | 5 | 1342 | BAB22812.1 | |
| AK003490.1 | 5 | 1342 | BAB22818.1 | |
| AK156619.1 | 5 | 1342 | BAE33778.1 | |
| AK157518.1 | 5 | 1342 | BAE34111.1 | |
| AK159385.1 | 5 | 1375 | BAE35040.1 | |
| BC043719.1 | 5 | 1391 | AAH43719.1 |
More Information
Set Membership: |
|

Genomic Map