Assay Details
Target Gene Details
Entrez Gene ID: | 72061 |
Gene Name: | RIKEN cDNA 2010111I01 gene |
Gene Aliases: |
2300006M17Rik, AP-O, Aopep, mir-23b, mir-24-1, mir-27b |
Location: |
Chr.13:62964893-63326096 on Build GRCm38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2010111I01Rik | NR_110521.1 | |||
| AK076298.1 | BAC36293.1 |
Target Gene Details
Entrez Gene ID: | 14088 |
Gene Name: | Fanconi anemia, complementation group C |
Gene Aliases: |
Facc, mir-3074-1 |
Location: |
Chr.13:63301134-63497290 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fancc | NM_001042673.2 | NP_001036138.1 | ||
| NM_001282942.1 | 14 | 2102 | NP_001269871.1 | |
| NM_007985.3 | 15 | 2299 | NP_032011.2 | |
| AK035488.1 | BAC29076.1 | |||
| AK042226.1 | ||||
| AK147025.1 | 15 | 2279 | BAE27618.1 | |
| BC038609.1 | 14 | 2084 | AAH38609.1 | |
| L08266.1 | 16 | 2379 | AAA37590.1 |
More Information
Set Membership: |
|

Genomic Map