Assay Details
Target Gene Details
Entrez Gene ID: | 69955 |
Gene Name: | phenylalanine-tRNA synthetase 2 (mitochondrial) |
Gene Aliases: |
Fars1, pheRS |
Location: |
Chr.13:36117643-36537595 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fars2 | NM_001039189.2 | NP_001034278.1 | ||
| NM_024274.3 | 2 | 884 | NP_077236.1 | |
| AK013211.1 | 2 | 623 | BAB28715.1 | |
| AK020164.1 | ||||
| AK076259.1 | 2 | 668 | ||
| AK170986.1 | 2 | 667 | BAE42160.1 | |
| AK189794.1 | ||||
| BC002147.1 | 2 | 654 | AAH02147.1 |
More Information
Set Membership: |
|

Genomic Map