Assay Details
Target Gene Details
Entrez Gene ID: | 14121 |
Gene Name: | fructose bisphosphatase 1 |
Gene Aliases: |
Fbp-2, Fbp2, Fbp3 |
Location: |
Chr.13:62864753-62888296 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbp1 | NM_019395.3 | 1 | 171 | NP_062268.1 |
| AJ132693.1 | 1 | 125 | CAB65244.1 | |
| AK002216.1 | BAB21941.1 | |||
| AK149527.1 | 1 | 52 | BAE28940.1 | |
| BC011480.1 | AAH11480.1 | |||
| BC051392.1 | AAH51392.1 |
More Information
Set Membership: |
|

Genomic Map