Assay Details
Target Gene Details
Entrez Gene ID: | 18814 |
Gene Name: | prolactin family 7, subfamily d, member 1 |
Gene Aliases: |
PLF-RP, PRP, Plfr |
Location: |
Chr.13:27708998-27716748 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prl7d1 | NM_011120.2 | 6 | 789 | NP_035250.1 |
| AK005421.1 | 6 | 786 | BAB24015.1 | |
| AK005557.1 | 6 | 790 | ||
| AK019461.1 | 6 | 789 | BAB31733.1 | |
| BC099470.1 | 6 | 778 | AAH99470.1 | |
| X02594.1 | 6 | 789 | CAA26437.1 |
More Information
Set Membership: |
|

Genomic Map