Assay Details
Target Gene Details
Entrez Gene ID: | 81003 |
Gene Name: | tripartite motif-containing 23 |
Gene Aliases: |
6330516O20Rik, AI450195, Arfd1 |
Location: |
Chr.13:104178774-104203421 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim23 | NM_030731.3 | 3 | 2344 | NP_109656.1 |
| AK029533.1 | 4 | 492 | ||
| AK030843.1 | 4 | 492 | BAC27156.1 | |
| AK030862.1 | 3 | 2345 | BAC27160.1 | |
| AK036210.1 | 4 | 493 | ||
| AK039280.1 | 4 | 504 | BAC30304.1 | |
| AK042070.1 | 4 | 495 | BAC31152.1 | |
| AK088922.1 | 4 | 492 | BAC40654.1 | |
| BC056390.1 | 4 | 485 | AAH56390.1 | |
| BC059017.1 | 4 | 485 | AAH59017.1 |
More Information
Set Membership: |
|

Genomic Map