Assay Details
Target Gene Details
Entrez Gene ID: | 195733 |
Gene Name: | grainyhead-like 1 (Drosophila) |
Gene Aliases: |
LBP-32, MGR, Tcfcp2l2 |
Location: |
Chr.12:24572230-24617391 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grhl1 | NM_001161406.1 | 16 | 2417 | NP_001154878.1 |
| NM_145890.2 | 14 | 2049 | NP_665897.2 | |
| AF411211.1 | 14 | 2049 | AAM22617.1 | |
| AF411212.1 | 16 | 2388 | AAM22618.1 | |
| AK076307.1 | 16 | 2403 | ||
| AK161048.1 | 16 | 2299 | BAE36170.1 | |
| BC013080.1 | 16 | 2357 | AAH13080.1 | |
| BC037233.1 | 16 | 2409 | AAH37233.1 |
More Information
Set Membership: |
|

Genomic Map