Assay Details
Target Gene Details
Entrez Gene ID: | 19819 |
Gene Name: | ribonuclease H1 |
Gene Aliases: |
- |
Location: |
Chr.12:28649602-28659591 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnaseh1 | NM_001286865.1 | 8 | 1153 | NP_001273794.1 |
| NM_011275.3 | 8 | 1153 | NP_035405.2 | |
| NR_104608.1 | 7 | 1094 | ||
| AF048993.1 | 8 | 1114 | AAC78562.1 | |
| AK011680.1 | 7 | 1068 | ||
| AK088032.1 | 6 | 1668 | BAC40109.1 | |
| BC019411.1 | 8 | 1082 | AAH19411.1 | |
| BM943446.1 | ||||
| DV662103.1 | 2 | 371 |
More Information
Set Membership: |
|

Genomic Map