Assay Details
Target Gene Details
Entrez Gene ID: | 15220 |
Gene Name: | forkhead box Q1 |
Gene Aliases: |
HFH-1, Hfh1, Hfh1l, sa |
Location: |
Chr.13:31558170-31560976 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxq1 | NM_008239.4 | 1 | 469 | NP_032265.3 |
| AK147202.1 | 1 | 475 | BAE27760.1 | |
| BC047155.1 | 1 | 202 | AAH47155.1 |
More Information
Set Membership: |
|

Genomic Map