Assay Details
Target Gene Details
Entrez Gene ID: | 11607 |
Gene Name: | angiotensin II receptor, type 1a |
Gene Aliases: |
1810074K20Rik, AG2S, AI551199, AT1, AT1a, AT2R1, AT2R1A, Agtr-1a, Agtr1, Angtr-1a |
Location: |
Chr.13:30336356-30382867 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agtr1a | NM_177322.3 | 3 | 1628 | NP_796296.1 |
| BC036175.1 | 3 | 1527 | AAH36175.1 |
More Information
Set Membership: |
|

Genomic Map