Assay Details
Target Gene Details
Entrez Gene ID: | 217866 |
Gene Name: | CDC42 binding protein kinase beta |
Gene Aliases: |
MRCKb |
Location: |
Chr.12:111292972-111377718 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc42bpb | NM_183016.2 | 30 | 4764 | NP_898837.2 |
| AK173103.1 | 17 | 2436 | BAD32381.1 | |
| AY277589.1 | 30 | 4405 | AAP34402.1 | |
| BC049921.1 | 5 | 905 | AAH49921.1 | |
| BC138035.1 | 30 | 4391 | ||
| BC138037.1 | 30 | 4391 |
More Information
Set Membership: |
|

Genomic Map