Assay Details
Target Gene Details
Entrez Gene ID: | 20259 |
Gene Name: | scinderin |
Gene Aliases: |
AW545522, adseverin |
Location: |
Chr.12:40059769-40134228 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scin | NM_001146196.1 | 16 | 2288 | NP_001139668.1 |
| NM_009132.2 | 14 | 1988 | NP_033158.2 | |
| AK133484.1 | 16 | 2244 | BAE21681.1 | |
| AK136080.1 | 14 | 1830 | BAE22809.1 | |
| AK167739.1 | 16 | 2243 | ||
| AK173295.1 | 16 | 2250 | BAD32573.1 | |
| BC063328.1 | 16 | 2243 | AAH63328.1 | |
| U04354.1 | 16 | 2243 | AAB61682.1 | |
| Y13971.1 | 14 | 1943 | CAA74304.1 |
More Information
Set Membership: |
|

Genomic Map