Assay Details
Target Gene Details
Entrez Gene ID: | 12950 |
Gene Name: | hyaluronan and proteoglycan link protein 1 |
Gene Aliases: |
BB099155, CLP, Crtl1, Crtl1l, LP, LP-1 |
Location: |
Chr.13:89540529-89611832 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hapln1 | NM_013500.4 | 3 | 653 | NP_038528.3 |
| AK003582.1 | 3 | 432 | ||
| AK032750.1 | 3 | 654 | BAC28007.1 | |
| AK048107.1 | 3 | 437 | BAC33244.1 | |
| BC066853.1 | 4 | 500 | AAH66853.1 |
More Information
Set Membership: |
|

Genomic Map