Assay Details
Target Gene Details
Entrez Gene ID: | 67295 |
Gene Name: | RAB3C, member RAS oncogene family |
Gene Aliases: |
2700062I01Rik, 3110015B08Rik, 3110037E15Rik, AI850886 |
Location: |
Chr.13:110054187-110281041 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab3c | NM_023852.5 | 5 | 7196 | NP_076341.1 |

Genomic Map