Assay Details
Target Gene Details
Entrez Gene ID: | 238463 |
Gene Name: | tubulin, alpha-like 3 |
Gene Aliases: |
- |
Location: |
Chr.13:3924635-3935277 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubal3 | NM_001033879.3 | 4 | 1318 | NP_001029051.2 |
| AK135971.1 | 4 | 1319 | BAE22753.1 | |
| BC147507.1 | 4 | 1311 |
More Information
Set Membership: |
|

Genomic Map