Assay Details
Target Gene Details
Entrez Gene ID: | 83428 |
Gene Name: | urocortin 3 |
Gene Aliases: |
- |
Location: |
Chr.13:3940688-3945349 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucn3 | NM_031250.5 | 1 | 338 | NP_112540.2 |
| AK148451.1 | 1 | 339 | BAE28561.1 |

Genomic Map