Assay Details
Target Gene Details
Entrez Gene ID: | 68680 |
Gene Name: | fat storage-inducing transmembrane protein 1 |
Gene Aliases: |
1110028A07Rik, AV010547, Cg10671, Fit1 |
Location: |
Chr.14:55575674-55576952 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fitm1 | NM_026808.1 | 2 | 383 | NP_081084.1 |
| AB054000.1 | 2 | 383 | BAB47400.1 | |
| AK003939.1 | 2 | 272 | BAB23085.1 | |
| BC037188.1 | 2 | 404 | AAH37188.1 | |
| BC048823.1 | 2 | 181 | AAH48823.1 | |
| BC104408.1 | 2 | 383 | AAI04409.1 | |
| BC104409.2 | 2 | 383 | AAI04410.1 |

Genomic Map