Assay Details
Target Gene Details
Entrez Gene ID: | 27384 |
Gene Name: | aldo-keto reductase family 1, member C13 |
Gene Aliases: |
mKIAA4014 |
Location: |
Chr.13:4191184-4205604 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c13 | NM_013778.2 | 9 | 1100 | NP_038806.2 |
| AB027125.1 | 9 | 1066 | BAA86850.1 | |
| AB097219.1 | 9 | 1066 | BAD02825.1 | |
| AK008949.1 | 9 | 1101 | BAB25986.1 | |
| AK220165.1 | 9 | 1093 | BAD90350.1 | |
| BC021937.1 | 9 | 1099 | AAH21937.1 |
More Information
Set Membership: |
|

Genomic Map