Assay Details
Target Gene Details
Entrez Gene ID: | 105387 |
Gene Name: | aldo-keto reductase family 1, member C14 |
Gene Aliases: |
9030611N15Rik, AW557061 |
Location: |
Chr.13:4048999-4090560 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c14 | NM_134072.1 | 9 | 1946 | NP_598833.1 |
| AK033537.1 | 9 | 1964 | BAC28346.1 | |
| AK136528.1 | 9 | 1963 | BAE23028.1 | |
| BC013482.1 | 9 | 1946 | AAH13482.1 | |
| BC026628.1 | 9 | 1945 | AAH26628.1 |
More Information
Set Membership: |
|

Genomic Map