Assay Details
Target Gene Details
Entrez Gene ID: | 12632 |
Gene Name: | cofilin 2, muscle |
Gene Aliases: |
- |
Location: |
Chr.12:54858815-54862877 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfl2 | NM_007688.2 | 4 | 2132 | NP_031714.1 |
| AK004659.1 | 4 | 2125 | ||
| AK020494.1 | 1 | 569 | ||
| AK162912.1 | 1 | 323 | ||
| BC007138.1 | 4 | 2075 | AAH07138.1 | |
| L29468.1 | 4 | 2132 | AAA37433.1 |
More Information
Set Membership: |
|

Genomic Map