Assay Details
Target Gene Details
Entrez Gene ID: | 12652 |
Gene Name: | chromogranin A |
Gene Aliases: |
ChrA |
Location: |
Chr.12:102554969-102565027 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chga | NM_007693.1 | 6 | 637 | NP_031719.1 |
| AK144504.1 | 6 | 631 | BAE25920.1 | |
| BC026554.1 | 6 | 621 | AAH26554.1 | |
| M64278.1 | 6 | 637 | AAA37457.1 |

Genomic Map