Assay Details
Target Gene Details
Entrez Gene ID: | 623046 |
Gene Name: | fibrous sheath CABYR binding protein |
Gene Aliases: |
EG623046 |
Location: |
Chr.12:64471333-64474898 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fscb | NM_001163271.1 | 1 | 1195 | NP_001156743.1 |
| EF133693.1 | 1 | 987 |

Genomic Map