Assay Details
Target Gene Details
Entrez Gene ID: | 18094 |
Gene Name: | NK2 homeobox 9 |
Gene Aliases: |
Nkx-2.9, Nkx2-8, Nkx2.9, tinman |
Location: |
Chr.12:56611389-56614676 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-9 | NM_008701.2 | 1 | 121 | NP_032727.2 |
| BC150762.1 | 1 | 104 | ||
| Y15741.1 | 1 | 123 | CAA75751.1 |
More Information
Set Membership: |
|

Genomic Map