Assay Details
Target Gene Details
Entrez Gene ID: | 21809 |
Gene Name: | transforming growth factor, beta 3 |
Gene Aliases: |
Tgfb-3 |
Location: |
Chr.12:86056743-86079041 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb3 | NM_009368.3 | 1 | 254 | NP_033394.2 |
| AK132519.1 | 1 | 242 | BAE21215.1 | |
| AK145304.1 | 1 | 242 | BAE26357.1 | |
| AK148385.1 | 1 | 118 | BAE28523.1 | |
| BC108426.1 | 1 | 254 | AAI08427.1 |
More Information
Set Membership: |
|

Genomic Map