Assay Details
Target Gene Details
Entrez Gene ID: | 21809 |
Gene Name: | transforming growth factor, beta 3 |
Gene Aliases: |
Tgfb-3 |
Location: |
Chr.12:86056743-86079041 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb3 | NM_009368.3 | 7 | 2394 | NP_033394.2 |
| AK132519.1 | 7 | 2384 | BAE21215.1 | |
| AK145304.1 | 7 | 2379 | BAE26357.1 | |
| AK148385.1 | 7 | 2258 | BAE28523.1 | |
| AK161328.1 | 4 | 641 | BAE36326.1 | |
| AK162560.1 | 7 | 1158 | BAE36968.1 | |
| BC005513.1 | 7 | 1136 | AAH05513.1 | |
| BC014690.1 | 7 | 1673 | AAH14690.1 | |
| BC108426.1 | 7 | 2394 | AAI08427.1 | |
| M32745.1 | 7 | 1889 | AAA40422.1 |
More Information
Set Membership: |
|

Genomic Map