Assay Details
Target Gene Details
Entrez Gene ID: | 68581 |
Gene Name: | transmembrane emp24-like trafficking protein 10 (yeast) |
Gene Aliases: |
1110014C03Rik, Tmp21, p23, p24delta1 |
Location: |
Chr.12:85340614-85374717 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed10 | NM_026775.4 | 5 | 2546 | NP_081051.1 |
| BC006774.1 | 3 | 2201 | AAH06774.1 |
More Information
Set Membership: |
|

Genomic Map