Assay Details
Target Gene Details
Entrez Gene ID: | 382620 |
Gene Name: | transmembrane emp24 domain containing 8 |
Gene Aliases: |
6430595O10Rik, AI447224, Gm1184, Mem1 |
Location: |
Chr.12:87166242-87200229 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed8 | NM_001033475.3 | 5 | 538 | NP_001028647.1 |
| AK147378.1 | 5 | 539 | BAE27873.1 | |
| BC150860.1 | 5 | 609 |

Genomic Map