Assay Details
Target Gene Details
Entrez Gene ID: | 83428 |
Gene Name: | urocortin 3 |
Gene Aliases: |
- |
Location: |
Chr.13:3940688-3945349 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucn3 | NM_031250.5 | 2 | 667 | NP_112540.2 |
| AK148451.1 | 2 | 668 | BAE28561.1 | |
| BC119433.1 | 1 | 152 | ||
| BC119434.1 | 1 | 149 |

Genomic Map