Assay Details
Target Gene Details
Entrez Gene ID: | 20184 |
Gene Name: | ubiquitin interaction motif containing 1 |
Gene Aliases: |
9430016E08Rik, D330018D10Rik, D630032M02Rik, RIP110, Rxrip110 |
Location: |
Chr.13:55027880-55100309 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uimc1 | NM_001293660.1 | 13 | 1955 | NP_001280589.1 |
| NM_011307.2 | 15 | 2793 | NP_035437.1 | |
| NR_121592.1 | 14 | 2712 | ||
| AK052704.1 | 15 | 2794 | BAC35106.1 |

Genomic Map