Assay Details
Target Gene Details
Entrez Gene ID: | 66694 |
Gene Name: | ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1 |
Gene Aliases: |
4430402G14Rik, AI875505 |
Location: |
Chr.13:30540312-30545316 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrfs1 | NM_025710.2 | 1 | 192 | NP_079986.1 |
| AK003966.1 | 1 | 155 | BAB23097.1 | |
| AK012180.1 | 1 | 143 | BAB28081.1 | |
| AK014470.1 | 1 | 195 | BAB29374.1 | |
| AK152391.1 | 1 | 204 | BAE31179.1 | |
| AK153139.1 | 1 | 179 | BAE31751.1 | |
| BC019934.1 | 1 | 199 | AAH19934.1 |

Genomic Map