Assay Details
Target Gene Details
Entrez Gene ID: | 66694 |
Gene Name: | ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1 |
Gene Aliases: |
4430402G14Rik, AI875505 |
Location: |
Chr.13:30540312-30545316 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uqcrfs1 | NM_025710.2 | 2 | 626 | NP_079986.1 |
| AK003966.1 | 2 | 589 | BAB23097.1 | |
| AK012180.1 | 2 | 577 | BAB28081.1 | |
| AK014470.1 | 2 | 629 | BAB29374.1 | |
| AK152391.1 | 2 | 638 | BAE31179.1 | |
| AK153139.1 | 2 | 613 | BAE31751.1 | |
| AK180820.1 | 1 | 162 | ||
| AK191429.1 | 1 | 135 | ||
| AK197173.1 | 1 | 109 | ||
| AK210928.1 | 1 | 111 | ||
| AK212564.1 | 1 | 123 | ||
| AK216624.1 | 1 | 123 | ||
| AK217705.1 | 1 | 123 | ||
| BC019934.1 | 2 | 633 | AAH19934.1 |

Genomic Map