Assay Details
Target Gene Details
Entrez Gene ID: | 14365 |
Gene Name: | frizzled homolog 3 (Drosophila) |
Gene Aliases: |
AU020229, D930050A07Rik, Fz3 |
Location: |
Chr.14:65192441-65262463 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fzd3 | NM_021458.2 | 4 | 1027 | NP_067433.1 |
| AK134261.1 | 4 | 963 | BAE22071.1 | |
| AK160554.1 | 4 | 986 | BAE35870.1 | |
| AK207512.1 | ||||
| U43205.1 | 3 | 876 | AAC52429.1 |
More Information
Set Membership: |
|

Genomic Map