Assay Details
Target Gene Details
Entrez Gene ID: | 18217 |
Gene Name: | neurotensin receptor 2 |
Gene Aliases: |
NT2R, NTRL |
Location: |
Chr.12:16653452-16660236 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntsr2 | NM_008747.2 | 4 | 1279 | NP_032773.2 |
| AB041826.1 | 4 | 1255 | BAB79256.1 | |
| AB041827.1 | 4 | 1255 | BAB79257.1 | |
| AK005368.1 | 4 | 936 | ||
| AK081073.1 | 3 | 2507 | ||
| AK158943.1 | 4 | 1279 | BAE34736.1 | |
| BB264011.1 | ||||
| BC141019.1 | 4 | 1274 | ||
| U51908.1 | 4 | 1278 | AAB17285.1 |
More Information
Set Membership: |
|

Genomic Map