Assay Details
Target Gene Details
Entrez Gene ID: | 238257 |
Gene Name: | transmembrane protein 30B |
Gene Aliases: |
9130011B11Rik |
Location: |
Chr.12:73543114-73546395 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem30b | NM_178715.3 | 1 | 1094 | NP_848830.1 |
| AK030302.1 | 1 | 1089 | BAC26887.1 | |
| AK033580.1 | 1 | 1095 | BAC28372.1 | |
| AK037246.1 | 1 | 1092 | BAC29774.1 | |
| AK169014.1 | 1 | 1130 | BAE40811.1 | |
| BC119224.1 | 1 | 1087 | ||
| BC119226.1 | 1 | 1087 |

Genomic Map