Assay Details
Target Gene Details
Entrez Gene ID: | 544922 |
Gene Name: | zinc finger with KRAB and SCAN domains 4 |
Gene Aliases: |
AI841875, Gm11272, OTTMUSG00000000421 |
Location: |
Chr.13:21478849-21485507 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan4 | NM_001039115.2 | 1 | 250 | NP_001034204.1 |
| BC141222.1 | 1 | 128 | ||
| BU148250.1 | 1 | 192 | ||
| CJ065384.1 | 1 | 252 |
More Information
Set Membership: |
|

Genomic Map