Assay Details
Target Gene Details
Entrez Gene ID: | 319178 |
Gene Name: | histone cluster 1, H2bb |
Gene Aliases: |
H2b-143 |
Location: |
Chr.13:23746734-23747223 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hist1h2bb | NM_175664.3 | 1 | 84 | NP_783595.1 |
| BC116853.1 | 1 | 84 | ||
| BC116857.1 | 1 | 84 |
More Information
Set Membership: |
|

Genomic Map