Assay Details
Target Gene Details
Entrez Gene ID: | 51791 |
Gene Name: | regulator of G-protein signaling 14 |
Gene Aliases: |
RPIP1 |
Location: |
Chr.13:55369712-55384691 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs14 | NM_016758.3 | 15 | 2242 | NP_058038.2 |
| AK002891.1 | 10 | 1731 | BAB22436.1 | |
| AK154944.1 | 14 | 2063 | ||
| BC030321.1 | 15 | 2214 | AAH30321.1 | |
| U85055.1 | 15 | 2082 | AAB41893.1 |

Genomic Map