Assay Details
Target Gene Details
Entrez Gene ID: | 630952 |
Gene Name: | angiogenin, ribonuclease A family, member 6 |
Gene Aliases: |
EG630952 |
Location: |
Chr.14:44001664-44011488 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ang6 | NM_001011876.2 | 2 | 273 | NP_001011876.1 |

Genomic Map