Assay Details
Target Gene Details
Entrez Gene ID: | 56419 |
Gene Name: | diaphanous homolog 3 (Drosophila) |
Gene Aliases: |
4930417P13Rik, Dia2, Diaph3, Drf3, mDia2, mKIAA4117, p134MDia2 |
Location: |
Chr.14:86655361-87141248 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Diap3 | NM_019670.1 | 28 | 3345 | NP_062644.1 |
| AF094519.1 | 28 | 3345 | AAC71771.1 | |
| AK015162.1 | 4 | 357 | ||
| AK220493.1 | 28 | 3409 | BAD90291.1 | |
| BC028920.1 | 21 | 2646 | AAH28920.1 | |
| BC085191.1 | 28 | 3455 | ||
| BC086779.1 | 28 | 3420 | AAH86779.1 |

Genomic Map