Assay Details
Target Gene Details
Entrez Gene ID: | 14560 |
Gene Name: | growth differentiation factor 10 |
Gene Aliases: |
Bmp3b |
Location: |
Chr.14:33923587-33935285 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf10 | NM_145741.2 | 1 | 205 | NP_665684.2 |
| BC058358.1 | 1 | 205 | AAH58358.1 | |
| L42114.1 | AAL77613.1 | |||
| S82648.1 | AAB46753.1 |
More Information
Set Membership: |
|

Genomic Map