Assay Details
Target Gene Details
Entrez Gene ID: | 11881 |
Gene Name: | arylsulfatase B |
Gene Aliases: |
1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1 |
Location: |
Chr.13:93771023-93943016 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsb | NM_009712.3 | 8 | 3144 | NP_033842.3 |
| AK003524.1 | 1 | 651 | BAB22836.1 |

Genomic Map