Assay Details
Target Gene Details
Entrez Gene ID: | 77041 |
Gene Name: | arylsulfatase K |
Gene Aliases: |
2810429K17Rik, 4833414G15Rik |
Location: |
Chr.13:76060422-76098667 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsk | NM_029847.4 | 4 | 813 | NP_084123.2 |
| AK013194.2 | 4 | 765 | BAB28703.1 | |
| AK019515.1 | 4 | 779 | BAB31772.1 | |
| AK031147.1 | 4 | 801 | BAC27279.1 | |
| AK032812.1 | 4 | 779 | BAC28035.1 | |
| AK033968.1 | 4 | 814 | ||
| AK035464.1 | 4 | 734 | BAC29070.1 | |
| AK039765.1 | 4 | 809 | BAC30444.1 | |
| AK046378.1 | 4 | 814 | BAC32696.1 | |
| AK083188.1 | 4 | 767 | BAC38801.1 | |
| AK084090.1 | 4 | 799 | BAC39115.1 | |
| AK086667.1 | 4 | 772 | BAC39714.1 | |
| AK139856.1 | 2 | 353 | ||
| AK144817.1 | 4 | 781 | BAE26079.1 | |
| AK165970.1 | 4 | 781 | BAE38491.1 | |
| BC046790.1 | 4 | 787 | ||
| BC058351.1 | 4 | 787 |

Genomic Map