Assay Details
Target Gene Details
Entrez Gene ID: | 105352 |
Gene Name: | dual specificity phosphatase 22 |
Gene Aliases: |
JKAP, JSP1, LMW-DSP2, MKPX |
Location: |
Chr.13:30660059-30711232 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp22 | NM_001037955.4 | 7 | 1986 | NP_001033044.1 |
| NM_134068.3 | NP_598829.1 | |||
| AF237619.1 | AAK15038.1 | |||
| AK149363.1 | 7 | 1945 | BAE28836.1 | |
| BC108362.1 | 7 | 1936 | AAI08363.1 | |
| CX236611.1 |
More Information
Set Membership: |
|

Genomic Map