Assay Details
Target Gene Details
Entrez Gene ID: | 15902 |
Gene Name: | inhibitor of DNA binding 2 |
Gene Aliases: |
AI255428, C78922, Idb2, bHLHb26 |
Location: |
Chr.12:25093799-25096092 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Id2 | NM_010496.3 | 3 | 881 | NP_034626.1 |
| AK003222.1 | 3 | 878 | BAB22651.1 | |
| AK135547.1 | 3 | 883 | BAE22577.1 | |
| AK146346.1 | 3 | 881 | BAE27099.1 | |
| AK146606.1 | 3 | 884 | BAE27298.1 | |
| BC006921.1 | 3 | 858 | AAH06921.1 | |
| BC053699.1 | 3 | 868 | AAH53699.1 | |
| BQ044638.1 | 1 | 142 | ||
| M69293.1 | 3 | 875 | AAA79771.1 |
More Information
Set Membership: |
|

Genomic Map