Assay Details
Target Gene Details
Entrez Gene ID: | 192657 |
Gene Name: | elongation factor RNA polymerase II 2 |
Gene Aliases: |
- |
Location: |
Chr.13:75707484-75772361 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ell2 | NM_138953.2 | 1 | 57 | NP_620403.2 |
| AK140906.1 | BAE24514.1 | |||
| AK144197.1 | ||||
| AK144978.1 | ||||
| AK145868.1 | BAE26710.1 | |||
| AK154545.1 | BAE32669.1 | |||
| AK156164.1 | ||||
| BC151188.1 |
More Information
Set Membership: |
|

Genomic Map