Assay Details
Target Gene Details
Entrez Gene ID: | 140580 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
6330578D22Rik, C230095H21Rik, CED-12 |
Location: |
Chr.13:20090240-20608353 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elmo1 | NM_080288.2 | 22 | 3273 | NP_525027.1 |
| NM_198093.3 | 7 | 1796 | NP_932761.1 | |
| AK049051.1 | 1 | 444 | ||
| AK220253.1 | 22 | 3161 | BAD90178.1 | |
| BC024727.1 | 7 | 1693 | AAH24727.1 | |
| BC031782.1 | 11 | 2011 | AAH31782.1 |
More Information
Set Membership: |
|

Genomic Map