Assay Details
Target Gene Details
Entrez Gene ID: | 386753 |
Gene Name: | DNA binding protein with his-thr domain |
Gene Aliases: |
Spot2 |
Location: |
Chr.12:74297474-74300468 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbpht2 | NM_198866.2 | 1 | 2487 | NP_942566.1 |

Genomic Map