Assay Details
Target Gene Details
Entrez Gene ID: | 56856 |
Gene Name: | insulinoma-associated 2 |
Gene Aliases: |
Mlt1 |
Location: |
Chr.12:55598917-55602018 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Insm2 | NM_020287.2 | 1 | 2590 | NP_064683.2 |

Genomic Map