Assay Details
Target Gene Details
Entrez Gene ID: | 260423 |
Gene Name: | histone cluster 1, H3f |
Gene Aliases: |
H3.2-221, Hist1 |
Location: |
Chr.13:23544052-23544954 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hist1h3f | NM_013548.4 | 1 | 336 | NP_038576.1 |
| BC101954.1 | 1 | 336 | AAI01955.1 | |
| BC101955.1 | 1 | 336 | AAI01956.1 | |
| BC101956.1 | 1 | 336 | AAI01957.1 | |
| BC103549.1 | 1 | 336 | AAI03550.1 |
More Information
Set Membership: |
|

Genomic Map